Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

E1618-97 Test Mix, Restek&Trade;
SDP

Catalog No. 06802002 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06-802-002 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06-802-002 Supplier Restek Corporation Supplier No. 31613
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Used for underground storage tank monitoring

Components:

  • (C6) n-Hexane (110-54-3)
  • (C8) n-Octane (111-65-9)
  • (C10) n-Decane (124-18-5)
  • (C12) n-Dodecane (112-40-3)
  • (C14) n-Tetradecane (629-59-4)
  • (C16) n-Hexadecane (544-76-3)
  • (C18) n-Octadecane (593-45-3)
  • (C20) n-Eicosane (112-95-8)
  • 2-Ethyltoluene (611-14-3)
  • 3-Ethyltoluene (620-14-4)
  • Toluene (108-88-3)
  • 1,2,4-Trimethylbenzene (95-63-6)
  • p-Xylene (106-42-3)

Specifications

Packaging Ampul
Concentration or Composition Notes Components in this mix (0.5 μL/mL or 0.05% volume/volume each) are at 10X the concentration of the final test solution
Ignition Point 556 deg C
Odor Strong
Vapor Density 2.93 (Air = 1)
Specific Gravity 1.3254 - 1.3258 g/cm3 at 20°C
Molecular Formula CH2Cl2
Concentration 10x (Concentration of the final test solution specified in ASTM 1618 and ASTM 1387)
MDL Number MFCD00000881
Color Colorless
Melting Point -96.7°C
Solubility Moderately soluble
Type Multi-component Standard
InChI Key YMWUJEATGCHHMB-UHFFFAOYSA-N
SMILES ClCCl
IUPAC Name dichloromethane
Molecular Weight (g/mol) 84.93
Quantity 1 mL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.