Learn More
Quality Biological Inc Medium 199 (E-199) without L-Glutamine and Phenol Red 500mL
SDP
Supplier:  Quality Biological Inc 112254101
Encompass_Preferred
This item has a minimum qty of 50 per supplier requirements.
One of the best known media in the history of cell culture Morgan Morton and Parkers Medium 199 was the first really successful attempt to develop a totally defined medium. While the developers succeeded in obtaining short term growth of explanted tissue they found that supplementation with serum was necessary for long term cultivation. This medium has been used for many different types of cells especially non-transformed cells and proved particularly useful in polio vaccine prod
Catalog No. 50-220-9741
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.