Learn More
Abcam E2-Ubiquitin Conjugation Kit, Enzyme Activity Assay, 50 Test
Description
- Assay type: Enzyme activity
- Sample types: Purified protein
- Cited in: > 6 publications
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | E2-Ubiquitin |
| No. of Reactions | 50 Tests |
| Format | Kit |
| Product Type | Western Blot Kit |
| Sample Type | Purified protein |
| Quantity | 50 Tests |
| For Use With (Application) | Functional studies, Western blot |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.