Learn More
Diagnostic Biosystems E2F-1 TRANSCRIPTION FACTOR E2F-1 Transcription Factor
Supplier:  Diagnostic Biosystems MOB38101
Encompass_Preferred
This antibody reacts with a protein of 60 kDa. E2Fs are DNAbinding proteins, which associate with negative regulators, such as the retinoblastoma p107 protein, resulting in an altered rate of gene transcription. E2F-1 is proposed to be involved in several cellular processes that range from tumor suppression, cell progression and oncogenesis. E2F-1 over expression can also drive cells into apoptosis.
Catalog No. 50-238-5471
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.