Learn More
Abcam E6 Berbamine, calmodulin (CaM) antagonist, 10MG
Supplier:  Abcam AB14151910MG
Encompass_Preferred
MW 757.8 Da, Purity >98%. Selective, cell-permeable CaM antagonist. Berbamine derivative. Inhibits calmodulin-dependent MYLK and PDE I (IC₅₀ values are 8 and 0.53 μM for MYLK and PDE1 respectively).
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6534