Learn More
Alkali Scientific Alkali Scientific™ Vornado™ BV101 Mini Vortex Mixer, 115V, Green
Supplier: Alkali Scientific RS101G
Powerful vortexing for tubes up to 50ml Inst-Touch™ pressure activated operation “Mini Sized”, less than 4 in. wide Available in 5 colors Powerful is the best word to describe this Vornado™ Mini Vortex Mixer. (Alkali Scientific™ BV101) With a 4 mm orbit and fixed speed of 2800rpm, the Vornado instantly vortexes even the largest samples, including nearly full 50 ml tubes. Despite its powerful motor, the Vornado has a footprint of less than 4×4 inches, allowing it to fit on even the most crowded bench. lnsta-Touch operation means that the motor is automatically activated with slight pressure on the center cup head. Operation is smooth, quiet and efficient. When pressure is removed, operation immediately ceases. The Vornado’s unique head design prevents liquids from entering the housing,prolonging motor life. The unit is available with five different cup head colors.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.