Learn More
Description
Provide more space to store hazardous material in your work environment. Eagle Floor Model Corrosives Cabinet can be used for storing containers up to 60 gallons in capacity.
- Suitable for storage of flammable and nonflammable acids and corrosive liquids
- Rugged 18-gauge steel cabinets coated with tough blue epoxy powder on interior and exterior surfaces, including shelves, for extra protection against corrosive acids and caustics
- Coated surfaces are superior to epoxy- or enamel-painted surfaces in case of spill or splash
- Doors clearly labeled CAUTION—CORROSIVES
- High-density polyethylene trays for shelves and bottom with drip edge contains liquid spills; removable for easy cleaning
Certifications
FM approved.
Compliance
Meet NFPA Code 30 and OSHA requirements.
Specifications
Specifications
| Dimensions (D x W x H) Interior | 15 x 40 x 41 in. |
| Product Type | Corrosives Cabinet |
| No. of Shelves | 1 |
| Width (Metric) Interior | 101.6 cm |
| Depth (Metric) Exterior | 45.7 cm |
| Height (Metric) Interior | 104.1 cm |
| Weight (Metric) | 115 kg |
| No. of Doors | 2 |
| Height (Metric) Exterior | 112 cm |
| Depth (Metric) Interior | 38 cm |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
