Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EAP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA530626
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA530626 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA530626 Supplier Invitrogen™ Supplier No. PA530626
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: A549, HeLa, HepG2, HCT116. Predicted reactivity: Pig (85%), Bovine (82%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes a member of a superfamily of divalent cation-dependent phosphodiesterases. The encoded protein associates with CD40, tumor necrosis factor (TNF) receptor-75 and TNF receptor associated factors (TRAFs), and inhibits nuclear factor-kappa-B activation. This protein has sequence and structural similarities with APE1 endonuclease, which is involved in both DNA repair and the activation of transcription factors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EAP2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.9 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene TDP2
Gene Accession No. O95551
Gene Alias 5'-Tyr-DNA phosphodiesterase; 5'-tyrosyl-DNA phosphodiesterase; AD022; AD-022; D13Ertd656e; dJ30M3.3; EAP2; EAPII; ETS1-associated protein 2; ETS1-associated protein II; hTDP2; MGC111021; MGC9099; RP1-30M3.3; Tdp2; TRAF and TNF receptor associated protein; TRAF and TNF receptor-associated protein; TTRAP; Tyr-DNA phosphodiesterase 2; tyrosyl-DNA phosphodiesterase 2; Tyrosyl-RNA phosphodiesterase; VPg unlinkase
Gene Symbols TDP2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human EAPII. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51567
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.