Learn More
Essendant EARPLUGSNO TCHSFTY CORD
Supplier:  Essendant MMMP2001
Encompass_Preferred
No-Touch Push-to-Fit Single-Use Earplugs combine the comfortable wearing experience of ultra-soft foam with a superior earplug design No-Touch Push-to-Fit format ensures the earplugs stay clean even when your hands or gloves are dirty Paddle-shaped livewire-stem grips allow you to insert and remove your earplugs without touching the plugs themselves Tapered bell-shaped extra-soft super-smooth foam tips accommodate a wide range of ear canal sizes and shapes-no rolling required All No-Touch Push-to-Fit Earplugs are compatible with the 3M E-A-Rfit Dual-Ear Validation System E-A-Rfit provides an accurate quantitative assessment of exactly how much noise-reduction a hearing protector will provide to any given worker Corded/Cordless Pair- Corded Shape- Bell Material(s)- Polyurethane Foam Thermoplastic Vinyl Noise Reduction Rate (NRR)- 29 dB
Catalog No. 50-000-31395
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.