Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Earth Sun Planetarium
SDPSCI_ED

Catalog No. S111028 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Grade:
6 to 14
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S111028 6 to 14
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S111028 Supplier United Scientific™ Supplier No. UNSLSY02
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This Earth Sun Planetarium is a dynamic visual depiction of what happens to sunlight once it gets to earth.

  • Demonstrate the rotational motion of Earth on its axis, its orbit around the Sun and the revolution of the moon around Earth with this high-quality model.
  • As the Sun casts a beam of light onto Earth (via an embedded LED), users can easily manipulate the positions of each body to examine how their dynamics correspond to seasons, eclipses, and lunar phases.

Specifications

Grade 6 to 14
Format Kit
Product Type Planetarium
Languages English
Subject Planetariums
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.