Learn More
StatLab EASY CUT DECAL. SOL 1GAL/EA
SDP
Supplier:  StatLab DCECDGAL
Encompass_Preferred
Easy Cut Decal Solution; 1 gal.; A medium strength decalcifier that renders excellent staining results; Perfect for routine use
Catalog No. NC9366318
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.