Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Easy Grip Tweezers
SCI_ED

Catalog No. S01717
Change view
Click to view available options
Length (Metric):
152 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
S01717 152 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
CHOKING HAZARD (1). Small parts. Not for children under 3 yrs.
Catalog No. S01717 Supplier Learning Resources, Inc Supplier No. LER1963
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CHOKING HAZARD (1). Small parts. Not for children under 3 yrs.

A perfect skill-building resource for teachers, specialists and occupational therapists.

Build skills as children use the tweezers to pick up and sort manipulatives, counters or any small objects.

  • Encourage proper thumb-forefinger pincer grasp and help to develop fine motor skills
  • Support early science explorations with true tweezer action Add excitement with six assorted colors: red, blue, green, yellow, orange and purple

Order Info

Grades PreK+. Tweezers measure 6 in. L (15.24cm); store in handy resealable bucket. Come in a pack of 12.

Specifications

Product Type Tweezers, Easy Grip
Length (English) 6 in.
Length (Metric) 152 mm
For Use With (Application) Primary science lessons, early science explorations and sorting activities
Includes Activity guide
Quantity Each
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.