Learn More
CHECKERS™ Easy Rider™ Speed Bump
Catalog No. 19085950 Shop All Justrite Safety Group Products
Change view
Click to view available options
Color:
Black, Yellow
Catalog No. Color
19-085-950 Black, Yellow
1 options
Catalog No. 19-085-950 Supplier CHECKERS™ Supplier No. 24111
Easy Rider™ Speed Bumps are maintenance-free and enhance parking lot safety
  • Made of 100% recycled rubber
  • Reduces speeds
  • High visibility at night
  • Simple installation in sections to create varying lengths
  • Will not warp, crack, chip or rot
  • Can be installed on any road surface
  • Resistant to ultraviolet light, moisture, oil, and extreme temperatures
  • Temporary or permanent installation – Relocatable when resurfacing / maintaining road surfaces;
  • Can be stored and re-used
  • Channeled bottom for cabling and piping
Specifications
Color Black, Yellow
Length (English) 4 in.
Packaging Quantity 1 Ea.
Material Plastic