Learn More
VAI Easy2Gown™ Mitcool 1700 Cleanroom Coveralls with Attached Hood and Boots
Catalog No. 19546550 Shop All VAI Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-546-550 3X-Large
19-546-545 Small
19-546-546 Medium
19-546-547 Large
19-546-548 X-Large
19-546-549 2X-Large
19-546-551 4X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 19-546-550 Supplier VAI Supplier No. 1800PEHBS1813
Add to Cart
Add to Cart
Cleanroom coveralls feature a proprietary fold that reduces the chance of the wearer contaminating the outside of the garment.
  • Bonded polypropylene with laminate
  • Sewn thumb loops to eliminate shifting
  • Piped seams with reinforced stitches
  • Excellent chemical resistance
  • Disposable
Specifications
Product Type Cleanroom Coveralls
Size 3X-Large
Color White
Disposable Single-use
Ankle Style Elastic
Cuff Style Elastic
Hood Type Attached, Open Face
Closure Type Zipper
Seam Type Piped, Stitched
Sterility Sterile
Model 1800P-EHB-S
For Use With (Application) Use in Class 100 (Grade A, ISO 5), Class 10,000 (Grade B, ISO 7) and in Class 100,000 (Grade C, ISO 8) areas in both aseptic and non-aseptic manufacturing and testing operations