Learn More
VAI Easy2Gown™ Mitcool 1700 Classic Cleanroom Coveralls
Catalog No. 19546423 Shop All VAI Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
6X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-546-423 2X-Large
19-546-425 4X-Large
19-546-421 Large
19-546-424 3X-Large
19-546-426 6X-Large
19-546-422 X-Large
19-546-419 Small
19-546-420 Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Catalog No. 19-546-423 Supplier VAI Supplier No. 1700PES17004
Add to Cart
Add to Cart
This item is not returnable. View return policy
Easy2Gown™ Mitcool 1700 Classic Cleanroom Coveralls feature a proprietary fold that reduces the chance of the individual contaminating the outside of the garment.
  • Bonded polypropylene with laminate
  • Sewn thumb loops to eliminate shifting
  • Piped seams with reinforced stitches
  • Excellent chemical resistance
  • Disposable
Specifications
Product Type Cleanroom Coveralls
Size 2X-Large
Material Polypropylene
Color White
Disposable Single-use
Ankle Style Elastic
Cuff Style Thumb Loop
Hood Type Unhooded
Seam Type Piped, Stitched
Sterility Sterile
Description Elastic wrist and ankle, double-stitched sewn thumb loops with bound edges
Model 1700P
For Use With (Application) Use in Class 100 (Grade A, ISO 5), Class 10,000 (Grade B, ISO 7) and in Class 100,000 (Grade C, ISO 8) areas in both aseptic and non-aseptic manufacturing and testing operations