Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Contec™ EasyCurve™ Mop Heads
BIOPHARMAGREENER_CHOICE

Catalog No. 19138940 Shop All Contec Inc. Products
Change view
Click to view available options
Material:
Knitted Polyester Microfiber, Hydrophilic Urethane Foam
Ribbed Polyester Microfiber, Hydrophilic Urethane Foam
Product Type:
Mop Head
Sealed Edge Mop Head
Sterility:
Non-sterile
Sterile
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material Product Type Sterility
19-138-940 Ribbed Polyester Microfiber, Hydrophilic Urethane Foam Sealed Edge Mop Head Non-sterile
19-159-243 Ribbed Polyester Microfiber, Hydrophilic Urethane Foam Mop Head Sterile
19-456-891 Knitted Polyester Microfiber, Hydrophilic Urethane Foam Sealed Edge Mop Head Sterile
19-159-242 Ribbed Polyester Microfiber, Hydrophilic Urethane Foam Mop Head Non-sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19-138-940 Supplier Contec™ Supplier No. EZSE0100
Add to Cart
Add to Cart

Lightweight mop for ceilings, walls, and floors. Choose from non-sterile or validated sterile versions. Mop heads are autoclavable. The EasyCurve™ Mop Heads use curved mop faces with pivoting connectors for easier surface contact at a variety of angles.

Contec's EasyCurve™ Mop was designed specifically for critical and controlled environments, now available with a ribbed microfiber face giving users smooth and easy mopping for better particle pick up and contamination removal. The combination of the curved mop face and the pivoting connector results in effortless surface contact and outstanding maneuverability, making the EasyCurve an ideal cleaning tool for ceilings, walls, and floors. All components of the EasyCurve Mop are autoclavable.

Features & Benefits

  • All components are autoclavable which is ideal for a wide range of cleaning applications in the most critical sterile environments
  • Lightweight mop head and handle significantly reduces operator fatigue
  • Single-use mop is economical and easy to install without tools
  • Curved mop and pivoting mop frame provide effortless contact with surface
  • Ribbed microfiber face provides better contaminant removal

Specifications

Product Type Sealed Edge Mop Head
Color Code White
Sterility Non-sterile
Autoclavable Autoclavable
Size 38 x 13 x 1 cm (15 x 5 x 0.375 in.)
Disposable Single-use
Quantity 2/Bag, 20 Bags/Cs.
For Use With (Equipment) EasyCurve Mop System
For Use With (Application) Designed for use in pharmaceutical and medical device manufacturing plants, hospitals, compounding pharmacies and other critical or controlled environments
Color-coded No
Material Ribbed Polyester Microfiber, Hydrophilic Urethane Foam
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.