Learn More
Vernier™ EasyLink
SDPSCI_ED
Catalog No. S15964ND Shop All Vernier Software Products
Change view
Click to view available options
For Use With (Application):
Educational use only
Catalog No. For Use With (Application)
S15964ND Educational use only
1 options
Catalog No. S15964ND Supplier Vernier™ Supplier No. EZLINK
Add to Cart
Add to Cart
Includes
Vernier EasyLink interface, Ordering Information, This product is discontinued and no longer available.
Inexpensive data collection direct to any TI-Nspire handheld or TI-84 Plus graphing calculator.
  • Support any one of over 45 Vernier sensors.
  • Use EasyLink to explore math and science concepts found in real-world phenomena such as light, pressure, force, and more
  • Connect a compatible sensor to EasyLink
  • Connect EasyLink to a TI-84 Plus or TI-Nspire handheld, and the Vernier data collection software launches automatically
Warranty and Services
5-year limited warranty
Specifications
Interface EasyLink Interface Included
For Use With (Application) Educational use only
For Use With (Equipment) TI-84 Plus graphing Calculator or TI-Nspire Handheld
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov