Learn More
Thermco Products EasyLog CO2 and Air Quality Data Logger
Catalog No. 04500345
Change view
Click to view available options
Software Type:
Web, Android, Apple IOS
Catalog No. Software Type
04-500-345 Web, Android, Apple IOS
1 options
Catalog No. 04-500-345 Supplier Thermco Products Supplier No. ELIOTCO2
Add to Cart
Add to Cart
EasyLog CO2 and Air Quality Data Logger, self-calibrating CO2 sensor, auto updates to cloud.
  • The EL-IOT-CO2 wireless data logger monitors and records CO2, temperature, and humidity levels, ideal for homes, workplaces, schools, and horticulture.
  • Continuously tracks parameters and displays current, minimum, and maximum values on a clear screen for quick air quality checks.
  • Custom high and low alarms can be set for each parameter to receive instant email or SMS alerts when thresholds are exceeded, ensuring prompt action.
  • Powered by USB with AA battery backup to ensure uninterrupted operation during brief power outages.
  • Measurement parameters include carbon dioxide (CO2), temperature, and humidity.
  • Provides real-time data with automatic cloud uploads for easy access to current and historical data.
  • EasyLog App allows users to view, analyze, and download data from any internet-enabled device for comprehensive air quality monitoring anytime, anywhere.
Specifications
Type Environmental Datalogger
Display Type LCD
Alarm Types E-mail, SMS
Software Type Web, Android, Apple IOS