Learn More
Thermco Products EasyLog High Accuracy Thermistor Probe Data Logger
Catalog No. 04500298
Change view
Click to view available options
For Use With (Equipment):
Data Loggers
Catalog No. For Use With (Equipment)
04-500-298 Data Loggers
1 options
Catalog No. 04-500-298 Supplier Thermco Products Supplier No. ELUSBTPLCDPLUS
Add to Cart
Add to Cart
Standalone data logger that records and displays over 32,000 temperature readings using the supplied thermistor probe.
  • Measures temperatures from -40°C to +125°C (-40°F to +257°F).
  • High-contrast LCD with push-button control allows viewing of recent highest and lowest readings.
  • Free EL-WIN-USB software enables easy configuration and data download via USB.
  • Supplied with a thermistor probe and long-life lithium battery.
  • Ideal for food service, manufacturing, and logistics applications.
  • Easy setup with plug-and-play configuration using free EasyLog software.
  • Extensive data analysis capabilities to visualize, print, and export data.
  • High accuracy ensures reliable temperature monitoring with large data capacity.
  • User-friendly display supports quick and efficient data review.
Specifications
Type Probe
For Use With (Equipment) Data Loggers