Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ EasyStrip™ Plus Tube Strip with Attached Ultra Clear Caps

Catalog No. AB2005 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
250 strips
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB2005 250 strips
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB2005 Supplier Thermo Scientific™ Supplier No. AB2005
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Includes

250 strips

The Thermo Scientific™ EasyStrip™ Plus Tube Strip with Attached Ultra Clear Caps is designed to reduce the risk of cross-contamination in qPCR assays.

The Thermo Scientific™ EasyStrip™ Plus Tube Strip with Attached Ultra Clear Caps is designed to reduce the risk of cross-contamination in qPCR assays. The tube strip is compatible with 0.2mL thermal cycler blocks and features individually attached optical caps for quick and easy access to individual samples. In addition, graduated 20μL markers on each tube enable visual volume verification and measurement of reagent and sample additions.

For tube strips with flat caps, please see Cat. No. AB-2000.

Features:
  • Avoid cross contamination with the individually attached caps
  • Verify tube reaction volumes with graduated 20μL markings on each tube
  • Track samples easily using the end tabs to label each strip
  • Identify individual wells quickly and prevent pipetting errors using the labels (A-H) on each tube cap and cap hinge (visible when cap is open and closed)
Applications
  • qPCR/Real-Time PCR

For Research Use Only. Not for use in diagnostic procedures.

Certifications

Certified human DNA, RNase and DNase free

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Certified DNA/RNase Free
Color Natural, Ultra Clear
Volume (Metric) Working 0.2 mL
Volume (Metric) 0.25 mL
Quantity 250 strips
For Use With (Equipment) Thermal Cyclers or Real-Time PCR Systems
Product Line EasyStrip™ 
Material Polypropylene
Shipping Condition Room Temperature
Closure Type Flat Cap
Format 250 Strip Tubes w/ Attached Caps
Product Type 8-strip Tube
Show More Show Less
What are the benefits of using EasyStrip Plus Tube Strip with Attached Ultra Clear Caps?

These new strips have new features including a graduated 20 µL measuring mark for visual verification and dual end tabs for better labeling and handling. In addition, the individual caps help prevent cross contamination and reduce sample evaporation.

Is the EasyStrip Plus Tube Strip with Attached Ultra Clear Caps DNase free?

Yes. The tubes are DNase/RNase/Human DNA free, as they are manufactured in a Class 100K ISO certified clean room UK production facility.

Which instruments are compatible with EasyStrip Plus Tube Strip with Attached Ultra Clear Caps?

Please refer to the table on Page 2 of this flyer (https://tools.thermofisher.com/content/sfs/brochures/easystrip-plus-tube-flyer.pdf) for compatibility information.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.