Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EBF2 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981629
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
50-198-1629 100 μL
1 options

Catalog No. 50-198-1629

Supplier: Bioss BS11740R

Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

EBF2 belongs to the conserved Olf/EBF family of helix-loop-helix transcription factors.

Specifications

Antigen EBF2
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene EBF2
Gene Accession No. Q9HAK2
Gene Alias COE2; Collier, Olf and EBF 2; D14Ggc1e; early B cell factor 2; Early B-cell factor 2; Ebf2; EBF-2; metencephalon-mesencephalnon-olfactory transcription factor 1; Metencephalon-mesencephalon-olfactory transcription factor 1; MET-mesencephalon-olfactory TF1; MET-mesencephalon-olfactory transcription factor 1; Mmot1; O/E-3; OE-3; Olf-1/EBF-like 3; Transcription factor COE2
Gene Symbols EBF2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human EBF2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64641
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.