Learn More
Aviva Systems Biology Corp Ebf4 Antibody - middle region (ARP37501_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37501_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Ebf4. It was validated on Western Blot. Ebf4,early B-cell factor 4,Ebf3; O/E-4; Olf-1; Ebf4,transcription factor COE4,transcription factor COE4,OE-4,EBF-4,Olf-1/EBF-like 4,OTTMUSP00000016528,OTTMUSP00000016532,early B-cell factor 3The immunogen is a synthetic peptide directed towards the middle region of human Ebf4. Alternate names: Ebf3, O/E-4, Olf-1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-284-644
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.