Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ eBioscience™ Essential Human Th1/Th17 Phenotyping Panel with Staining & Stimulation Reagents

Catalog No. A42926
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A42926 100 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A42926 Supplier Invitrogen™ Supplier No. A42926
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Invitrogen™ eBioscience™ Essential Human Th1/Th17 Phenotyping Panel with Staining and Stimulation Reagents is designed for flow cytometry-based identity testing of Type 1 and Type 17 T-helper cells

Phenotyping Panel with Staining and Stimulation Reagents leverages high quality eBioscience™ antibodies, cell stimulation reagents (induces cytokine production), and fixation/permeabilization reagents (for intracellular markers IfN-g and IL-17) and includes a ready-to-use protocol for experimental setup, gating strategy, and analysis.

Benefits include:

  • Proven performance―antibodies and supporting reagents designed for flow cytometry
  • Highly referenced antibodies―includes one of the most referenced clones for IL-17
  • Turnkey kit and protocol―contains reagents and standardized gating recommendations needed to perform flow cytometry characterization
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage
  • CD4, Ifn-g, IL-17a antibodies, store at 2°C to 8°CC
  • Cell stimulating cocktail, store at -5 to -30°C
  • Protein transport cocktail, store at -5 to -30°C
  • Intracellular fixation & permealization reagents, store at 2°C to 8°CC
Format Kit
Quantity 100 reactions
Detection Method Fluorescence
Shipping Condition Approved for shipment at Room Temperature or on Wet or Dry Ice
Product Line eBioscience
Product Type Phenotyping Panel

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.