Learn More
Invitrogen™ eBioscience™ Human Cell Surface Flow Cytometry Workflow Kit
Description
- Kit contains all the buffers, blockers, and beads needed to complete a standard flow cytometry experiment for staining cell surface targets on human cell suspensions
- When combined with experiment-specific antibodies, the reagents within this kit have been optimized to work together to detect cell surface targets by flow cytometry
- Effective compensation controls
- Fc receptor blocking
- Non-antibody, non-protein-based blocking solution for reducing background staining from fluorophores
- Antibody binding buffer minimizes non-specific binding of antibodies
- Fixation buffer for storage of stained samples
Kit Includes:
- UltraComp eBeads™ Plus Compensation Beads (01-3333-41)
- Fc Receptor Binding Inhibitor Polyclonal Antibody, eBioscience (14-9161-71)
- CellBlox Monocyte and Macrophage Blocking Buffer (B001T02F01)
- eBioscience Flow Cytometry Staining Buffer (00-4222-57)
- eBioscience IC Fixation Buffer (00-8222-49)
Specifications
Specifications
| Quantity | 1 Kit |
| For Use With (Application) | Flow Cytometry |