Learn More
Invitrogen™ eBioscience™ Human Intracellular (cytoplasmic/cytokine) Protein Flow Cytometry Workflow Kit
Catalog No. A53014
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 Kit
Catalog No. Quantity
A53014 1 Kit
1 options
Catalog No. A53014 Supplier Invitrogen™ Supplier No. A53014
Only null left
Add to Cart
Add to Cart
The eBioscience™ Human Intracellular (cytoplasmic/cytokine) Protein Flow Cytometry Workflow Kit contains all of the normal buffers, blockers, and beads needed to complete a standard flow cytometry experiment for staining intracellular cytoplasmic and cytokine targets in human cell suspensions.
  • Kit contains all of the normal buffers, blockers, and beads to complete a standard flow cytometry experiment for staining intracellular cytoplasmic and cytokine targets in human cell suspensions
  • When combined with experiment-specific antibodies, the reagents within this kit have been optimized to work together to detect human intracellular cytopasmic and cytokine targets by flow cytometry
  • Effective compensation controls
  • Fc receptor blocking
  • Non-antibody, non-protein based blocking solution for reducing background staining from fluorophores
  • Antibody binding buffer minimizes non-specific binding of antibodies
  • Fixation buffer for storage of stained samples
  • Activating cocktail to induce cytokine production
Kit includes
  • UltraComp eBeads™ Plus Compensation Beads (01-3333-41)
  • Fc Receptor Binding Inhibitor Polyclonal Antibody, eBioscience (14-9161-71)
  • CellBlox™ Blocking Buffer (B001T06F01)
  • eBioscience Intracellular Fixation & Permeabilization Buffer Set (88-8824-00)
  • eBioscience™ Flow Cytometry Staining Buffer (00-4222-57)
  • eBioscience Cell Stimulation Cocktail plus protein transport inhibitors, 500X (00-4975-93)
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 Kit
For Use With (Application) Flow Cytometry