Learn More
Invitrogen™ eBioscience™ Human Lysed Whole Blood Cell Flow Cytometry Workflow Kit
Description
- Kit contains all of the normal buffers, blockers, and beads to complete a standard flow cytometry experiment for staining cell surface targets on human lysed whole blood
- When combined with experiment-specific antibodies, the reagents within this kit have been optimized to work together to detect human blood cell surface reagents by flow cytometry
- Effective compensation controls
- FC receptor blocking
- Non-antibody, non-protein based blocking solution for reducing background staining from fluorophores
- Antibody binding buffer maximizes non-specific binding of antibodies
- Fixation buffer for storage of stained samples
Kit Includes
- UltraComp eBeads™ Plus Compensation Beads (01-3333-41)
- eBioscience 10X RBC Lysis Buffer, multi-species (00-4300-54)
- Fc Receptor Binding Inhibitor Polyclonal Antibody, eBioscience (14-9161-71)
- CellBlox™ Blocking Buffer (B001T06F01)
- eBioscience™ Flow Cytometry Staining Buffer (00-4222-57)
- eBioscience™ IC Fixation Buffer (00-8222-49)
Specifications
Specifications
| Quantity | 1 Kit |
| For Use With (Application) | Flow Cytometry |