Learn More
Invitrogen™ eBioscience™ Mouse Intracellular (cytoplasmic/cytokine) Protein Flow Cytometry Workflow Kit
Description
- Kit contains all of the normal buffers, blockers and beads to complete a standard flow cytometry experiment for staining intracellular cytoplasmic and cytokine targets in mouse cell suspensions
- When combined with experiment-specific antibodies, the reagents within this kit have been optimized to work together to detect mouse intracellular cytoplasmic and cytokine targets by flow cytometry
- Effective compensation controls
- Fc receptor blocking
- Non-antibody, non-protein based blocking solution for reducing background staining from fluorophores
- Antibody binding buffer minimizes non-specific binding of antibodies
- Fixation buffer for storage of stained samples
- Activating cocktail to induce cytokine production
Kit Includes
- UltraComp eBeads™ Plus Compensation Beads (01-3333-41)
- CD16/CD32 Monoclonal Antibody (93), eBioscience (14-0161-81)
- CellBlox™ Blocking Buffer (B001T06F01)
- eBioscience Intracellular Fixation & Permeabilization Buffer Set (88-8824-00)
- eBioscience™ Flow Cytometry Staining Buffer (00-4222-57)
- eBioscience Cell Stimulation Cocktail plus protein transport inhibitors, 500X (00-4975-93)
Specifications
Specifications
| Quantity | 1 Kit |
| For Use With (Application) | Flow Cytometry |