Learn More
Invitrogen™ eBioscience™ Mouse Intracellular (nuclear/transcription factor) Protein Flow Cytometry Workflow Kit
Description
- Kit contains all of the normal buffers, blockers, and beads to complete a standard flow cytometry experiment for staining intracellular nuclear and transcription factor targets in human cell suspensions
- When combined with experiment-specific antibodies, the reagents within this kit have been optimized to work together to detect mouse intracellular nuclear and transcription targets by flow cytometry
- Effective compensation controls
- Fc receptor blocking
- Non-antibody, non-protein based blocking solution for reducing background staining from fluorophores
- Antibody binding buffer which minimizes non-specific binding of antibodies
- Staining buffer optimized for transcription factors and nuclear proteins
Kit Includes
- UltraComp eBeads™ Plus Compensation Beads (01-3333-41)
- CD16/CD32 Monoclonal Antibody (93), eBioscience (14-0161-81)
- CellBlox™ Blocking Buffer (B001T06F01)
- eBioscience Foxp3/Transcription Factor Staining Buffer Set (00-5523-00)
- eBioscience™ Flow Cytometry Staining Buffer (00-4222-57)
Specifications
Specifications
| Quantity | 1 Kit |
| For Use With (Application) | Flow Cytometry |