Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ eBioscience™ Phalloidin eFluor™ 570

Catalog No. 41655905
Encompass_Preferred
Change view
Click to view available options
Quantity:
300 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
41-655-905 300 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 41-655-905 Supplier Invitrogen™ Supplier No. 41655905
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Phalloidin is a peptide toxin isolated from the Amanita phalloides mushroom. Phalloidin selectively binds F-actin and functions to stablilize actin polymers by inhibiting the dissociation of actin monomers from the filament ends.

Phalloidin is a peptide toxin isolated from the Amanita phalloides mushroom. Phalloidin selectively binds F-actin and functions to stablilize actin polymers by inhibiting the dissociation of actin monomers from the filament ends. Because F-actin polymers compose the cytoskeleton of almost all eukaryotic cells, phalloidin conjugates can be useful probes for visualizing the structure of live or fixed cells using microscopy. Phalloidin eFluor™ 570 can be used to label cells of multiple species.

Phalloidin eFluor™ 570 is supplied as a frozen liquid. To prepare the 1000X stock solution, thaw the vial, add 30 μL high-quality anhydrous DMSO, mix and briefly spin down. The resulting solution (phalloidin DMSO stock) should be aliquoted, protected from light and moisture, and stored at -20°C. To prepare a 1X working solution of phalloidin conjugate, add 1 μL of 1000X phalloidin DMSO stock solution to 1 mL of PBS containing 1% BSA.

Reported Applications

  • Immunocytochemistry
  • Immunohistochemical Staining
  • Microscopy

TRUSTED_SUSTAINABILITY

Specifications

Excitation Wavelength Range 555/570 nm
Dye Type eFluor 570
No. of Tests 300 Tests
Quantity 300 tests
Product Line eBioscience
Label Type eFluor
Product Type Phalloidin
Sub Cellular Localization Cytoskeleton
Can I combine Click-iT or Click-iT Plus reactions with phalloidin conjugates used for actin staining?

We do not recommend using phalloidin conjugates for staining actin in combination with traditional Click-iT or Click-iT Plus reactions since phalloidin is extremely sensitive to the presence of copper.

For staining actin in combination with traditional Click-iT or Click-iT Plus reactions, we recommend using anti-α-actin antibodies for staining actin in the cytoskeleton. You can find a list of our actin antibodies here.

Another option would be to use the Click-iT Plus Alexa Fluor Picolyl Azide Toolkit (Cat. Nos. C10641, C10642, C10643). These Click-iT Plus toolkits provide Copper and Copper protectant separately which makes it easier to titrate the copper concentration to obtain optimal labeling with minimal copper-mediated damage. You may need to optimize the click reaction with the lowest possible concentration of copper and then perform the phalloidin staining.

I am unable to analyze my cells stained with eFluor Organic Dyes today. What options do I have?

Our options will depend on the samples you are analyzing.

If cell viability is not critical, you can store your stained samples at 4 degrees C or on ice overnight in the dark and analyze the following day.

For samples stained with eFluor organic fluorochromes, we recommend that cells be suspended in 100 uL of Flow Cytometry Staining Buffer (Cat. No. 00-4222) and 100 uL of eBioscience IC Fixation Buffer (Cat. No. 00-8222); samples can be incubated for up to 3 days at 4 degrees C in the dark. Alternatively, the 1-step Fix/Lyse Solution (Cat. No. 00-5333) can be used. This is a great option when working with whole blood but also works for other cell types.

Can the eFluor Organic fluorochromes be used for intracellular staining?

Yes, the eFluor Organic fluorochromes can be used for intracellular staining. The eFluor organic fluorochromes maintain bright signal and require minimal changes in compensation when fixed with eBioscience IC Fixation Buffer (Cat. No. 00-8222-49) and Permeabilization Buffer (Cat. No. 00-8333-56) or 1-step Fix/Lyse Solution (Cat. No. 00-5333-54, 00-5333-57) (as compared to live cells).

What buffers are compatible with the eFluor Organic fluorochromes and eVolve QDots?

The eFluor Organic fluorochromes and eVolve QDots can be used with flow staining buffers containing PBS and protein.

What is the difference between eFluor Organic Dyes and eVolve Qdots?

The eFluor Organic Dyes (eFluor 450, APC-eFluor 780, PerCP-eFluor 710, eFluor 710) are conventional fluorochromes. In contrast, the eVolve line of products are Quantum dots.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.