Learn More
Enzo Life Sciences Ebola virus monoclonal antibody (FE25) (100µg)
SDP
Supplier:  Enzo Life Sciences ENZABS2670100
Encompass_Preferred
Clone: FE25. Isotype: IgG2a. Immunogen: Ebola virus (formalin inactivated, Zaire strain). Species reactivity: Ebola virus. Specificity: Recognizes ebola virus in human samples. Applications: ELISA, WB. Purity: Protein G affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 0.09% sodium azide.
Catalog No. 50-201-2812
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.