Learn More
Eisco™ Ebola Virus Model
SCI_ED
Catalog No. S27905 Shop All Eisco Products
Change view
Click to view available options
PVC
Catalog No.
S27905 PVC
1 options
Catalog No. S27905 Supplier Eisco™ Supplier No. ZM101816
Add to Cart
Add to Cart
Enlarged Ebola virus model effectively shows the various membrane layers and structure of the virus, including a partial sectional view depicting proteins.
  • Enlarged 100,000 times
  • Features various membrane layers and structure of the virus
  • Cut-away at one end exposes internal structures, major and minor matrix proteins, polymerase protein, and Ebola RNA
  • Made of durable polyvinyl chloride (PVC) plastic which is corrosion resistant, lightweight, and has high strength
  • Resists reactions with acids, gasoline, alcohol, and hydrocarbons
  • Vividly hand painted with eco safe paint
  • Supplied with English Key Card
Specifications
For Use With (Application) Virus Studies
Material PVC
Product Type Model