Learn More
Sino Biological EBOV EBOV-G PAb 20UL 40094-T46-20
Supplier:  Sino Biological 40094T4620
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Ebola virus EBOV (Subtype Sudan, strain Gulu) Glycoprotein / GP ( Catalog#40094-V08B1; YP_138523.1; Met1-Asn637). Ebola virus EBOV (Subtype Sudan, strain Gulu) Glycoprotein / GP specific IgG was purified by Ebola virus EBOV (Subtype Sudan, strain Gulu) Glycoprotein / GP affinity chromatography.
Catalog No. 50-241-1970
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.