Learn More
Invitrogen™ EBP50 Monoclonal Antibody (EBP-10)

Description
This antibody reacts with NHERF1/EBP50 phosphoprotein of 50 kDa, which serves as an adaptor and regulator protein. For use in Western blot applications, MA1-19291 should be placed in a vertical incubator for 60 minutes. A suggested positive control for this product is RAJI human lymphoma cell lysate. Western Blot: Reducing conditions.
Specifications
Specifications
| Antigen | EBP50 |
| Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | EBP-10 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 15mM sodium azide; pH 7.4 |
| Gene | SLC9A3R1 |
| Gene Accession No. | O14745 |
| Gene Alias | EBP50; EBP-50; ERM-binding phosphoprotein; ezrin-radixin-moesin binding phosphoprotein-50; ezrin-radixin-moesin-binding phosphoprotein 50; Na(+)/H(+) exchange regulatory cofactor NHE-RF1; Na+/H+ exchange regulatory co-factor; Nherf; NHE-RF; Nherf1; NHERF-1; NPHLOP2; Regulatory cofactor of Na(+)/H(+) exchanger; SLC9A3 regulator 1; Slc9a3r1; So; Sodium-hydrogen exchanger regulatory factor 1; solute carrier family 9 (sodium/hydrogen exchanger), isoform 3 regulator 1; solute carrier family 9 (sodium/hydrogen exchanger), isoform 3 regulatory factor 1; solute carrier family 9 (sodium/hydrogen exchanger), member 3 regulator 1; solute carrier family 9 isoform A3 regulatory factor 1; solute carrier family 9, subfamily A (NHE3, cation proton antiporter 3), member 3 regulator 1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.