Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Ebselen
GREENER_CHOICESDP

Catalog No. 524550
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
52-455-0 50 mg
52-451-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 52-455-0 Supplier Tocris Bioscience™ Supplier No. 5245/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Glutathione peroxidase mimic; peroxynitrite scavenger

Ebselen is a glutathione peroxidase mimic; peroxynitrite scavenger. Inhibits ferroptosis. Also inhibits lipoxygenase, cyclooxygenase, nitric oxide synthase, protein kinase C and H+/K+-ATPase activity. Inhibits the hepatic carcinogenic effects of aflatoxin B1. Antioxidant and anti-inflammatory. Selectively inhibits Gq protein signaling and enhances differentiation of brown adipocytes. Inhibits SARS-CoV-2 Mpro in vitro (IC50 = 0.67 μM in FRET assay)

Specifications

CAS 60940-34-3
Quantity 50 mg
Molecular Formula 2-Phenyl-1,2-benzisoselenazol-3(2 H)-one
Product Type Ebselen
Purity 98%
Molecular Weight (g/mol) M.W. (Observed): 274.18
Inhibitors Ebselen

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.