Learn More
BioVendor, LLC EBV EBNA-1, E. coli Recombinant Protein (0.1 mg). Epstein-Barr virus nuclear antigen 1.
SDP
Supplier:  BioVendor, LLC RD972344100
Encompass_Preferred
EBV EBNA-1 (Epstein-Barr virus nuclear antigen 1), E. coli Recombinant Protein, 0.1 mg. DATA: Total 242 AA. MW: 25.9 kDa. UniProtKB# P03211 (His407-Glu641). N-terminal His-tag (7 extra AA). PURITY >95%. ENDOTOXIN <1.0 EU/ug. FORM: Filtered 0.4 m, lyophilized from 0.5 mg/ml solution in PBS, pH 7.4. SUMMARY: EBNA1 plays an essential role in replication & partitioning of viral genomic DNA during latent infection. During this phase, the circular viral dsDNA undergoes replication once per cell cycle and is efficiently partitioned to the daughter cells. EBNA1 activates initiation of viral DNA replication by binding to specific sites in the viral latent origin of replication, oriP. Also, it governs the segregation of viral episomes by mediating their attachment to host cell metaphase chromosomes. Also activates transcription of several viral latency genes. Finally, it can counteract stabilization of host p53/TP53 by host USP7, thereby decreasing apoptosis & increasing host cell survival.
Catalog No. 50-206-9208
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.