Learn More
STEMCELL Technologies EBV (EBNA-1) Peptide Pool, Size: 1 Unit
SDP
Supplier:  STEMCELL Technologies 1000669
Encompass_Preferred
The EBV (EBNA-1) Peptide Pool is a lyophilized mixture of 158 peptides from Epstein-Barr nuclear antigen 1 (EBNA-1) of Epstein-Barr virus (EBV strain B95-8). EBNA-1 plays a critical role in the stable latent infection by EBV and may contribute to the survival of EBV-infected cells through its interaction with ubiquitin carboxyl-terminal hydrolase 7 (USP7). The pool consists of 15-mer peptides with 11-amino-acid overlaps that cover amino acids 1 - 641 on EBNA-1.
Catalog No. 50-239-0140
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.