Learn More
Eenzyme LLC EBV HHV-4 GH/GL/GP42 PRO 50UG
Supplier:  Eenzyme LLC HHV4GHL4223P
Encompass
C-terminal 6xHis-tagged recombinant EBV (or HHV-4) gH (aa 1-344)(GenBank accession#: P03231) in complex with gL (aa 25-137)(GenBank accession#: CAD53428) and gp42 (aa 31-223)(Genbank accession#: YP_401672) 50ug
Catalog No. NC2379654
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.