Learn More
Drg International Inc DRG EBV-VCA IgG ELISA RUO, 96 Well Kit
Supplier:  Drg International Inc EIA3475R
Encompass_Preferred
An enzyme immunoassay for the qualitative and semiquantitative determination of IgG-class antibodies to Epstein Barr Virus (EBV) -Viral Capsid Antigen (VCA) in serum and plasma. This product is for Research Use Only. The DRG Epstein Barr Virus (VCA) IgG ELISA Kit is a solid phase enzyme-linked immunosorbent assay (ELISA). Microtiter wells as a solid phase are coated with inactivated Epstein Barr Virus (VCA) antigen (rec. protein p18 + p23). Diluted specimens and ready-for-use controls are pipetted into these wells.
Catalog No. 50-194-7569
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.