Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ECEL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA527951
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA527951 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA527951 Supplier Invitrogen™ Supplier No. PA527951
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: ECEL1 tansfected 293T cell. Predicted reactivity: Mouse (95%), Rat (95%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes a member of the M13 family of endopeptidases. In general, M13 family members are zinc-containing type II integral-membrane proteins that are important regulators of neuropeptide and peptide hormone activity. This gene is expressed specifically in the central nervous system and its protein localizes predominately to the endoplasmic reticulum or, in trace amounts, to the cell surface. Disruption of this gene in mouse embryonic stem cells results in neonatal lethality due to respiratory failure shortly after birth. Based on the specific expression of this gene and the phenotype of the gene deficiency in mouse embryos, it is suggested that this protein plays a critical role in neural regulation of the respiratory system. This gene has multiple pseudogenes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ECEL1
Applications Western Blot
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene ECEL1
Gene Accession No. O95672
Gene Alias DA5D; damage induced neuronal endopeptidase; damage-induced neuronal endopeptidase; Dine; ECEL1; ECEX; endothelin converting enzyme like 1; endothelin converting enzyme-like 1; endothelin-converting enzyme-like 1; metallopeptidase; UNQ2431/PRO4991; X converting enzyme; XCE; Xce protein
Gene Symbols ECEL1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human ECEL1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9427
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.