Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EHHADH Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583586
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583586 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583586 Supplier Invitrogen™ Supplier No. PA583586
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ILGEGIAASP EHIDVVYLHG YGWPRHKGGP MFYASTVGLP TVLEKLQKYY RQNPDIPQLE PSDYLKKLAS QGNPPLKE Highest antigen sequence indentity to the following orthologs: Rat - 86%, Mouse - 85%.

As an L-hydroxy-specific enzyme, EHHADH (enoyl-CoA-hydratase:3-hydroxyacyl-CoA dehydrogenase), also known as Peroxisomal L-bifunctional enzyme, is a 723 amino acid protein has an essential tripeptide sequence on its carboxyl-terminus that is required for peroxisomal transport. EHHADH-null mice only exhibit a blunted peroxisome proliferative response when challenged with a peroxisome proliferator. Since there were no observed changes in lipid metabolism, this evidence suggests that enoyl-CoAs were diverted to the D-hydroxy-specific beta-oxidation system for metabolism.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EHHADH
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene EHHADH
Gene Accession No. Q08426
Gene Alias 1; 1300002P22Rik; 3,2-trans-enoyl-CoA isomerase; 3-hydroxyacyl-CoA dehydrogenase; ECHD; EHHADH; Enoyl-CoA hydratase/3,2-trans-enoyl-CoA isomerase; enoyl-CoA, hydratase/3-hydroxyacyl CoA dehydrogenase; enoyl-Coenzyme A, hydratase/3-hydroxyacyl Coenzyme A dehydrogenase; FRTS3; HD; L-3-hydroxyacyl-CoA dehydrogenase; LBFP; L-bifunctional enzyme; L-bifunctional protein, peroxisomal; Lbp; L-PBE; L-specific bifunctional protein; L-specific multifunctional beta-oxdiation protein; MEF; Mfe; Mfe1; MFP; MFP1; multifunctional enzyme type 1; Pbe; PBFE; perMFE-1; Peroxisomal bifunctional enzyme; peroxisomal bifunctional enzyme type 1; peroxisomal enoyl-CoA hydratase
Gene Symbols EHHADH
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human EHHADH. Recombinant protein control fragment (Product #RP-103761).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1962
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.