Learn More
Tocris Bioscience™ Echistatin, alpha1 isoform
GSA_VAGREENER_CHOICE
Catalog No. 3202100U
Change view
Click to view available options
Quantity:
100 μg
Catalog No. Quantity
32-021-00U 100 μg
1 options
This item is not returnable. View return policy
Catalog No. 32-021-00U Supplier Tocris Bioscience™ Supplier No. 3202/100U
Add to Cart
Add to Cart
This item is not returnable. View return policy
αVβ3 and glycoprotein IIb/IIIa (integrin αIIbβ3) inhibitor
Echistatin, α1 isoform is a potent irreversible αVβ3 integrin antagonist (Ki = 0.27 nM). Disrupts attachment of osteoclasts to bone and inhibits bone reabsorption (IC50 = 0.1 nM). Prevents ADP-induced platelet aggregation via inhibition of glycoprotein IIb/IIIa (GpIIb/IIIa, αIIbβ3) receptors (IC50 = 30 nM) in vitro.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.