Learn More
Mitegen IN35 Chilling Heating Incubator By EchoTherm, 27.5L Capacity, Fully Programmable Temperature Control
SDP
Supplier:  Mitegen TPSIN35
Encompass
The EchoTherm IN35 Chilling Heating Incubator provides precise and versatile temperature control for a wide range of laboratory applications. With its substantial 27.5-liter capacity, this incubator is ideal for maintaining stable conditions for sensitive samples, reagents, and cultures. Featuring fully programmable control, users can easily set and adjust heating and chilling cycles, ensuring optimal thermal environments for their experiments. This incubator delivers reliable performance for applications requiring temperatures both above and below ambient, such as enzyme kinetics, cell culture, and molecular biology assays. Its advanced temperature management capabilities contribute to enhanced reproducibility and accuracy in scientific research. Designed for ease of use and long-term durability, it offers a dependable solution for any laboratory demanding consistent and precise thermal regulation for critical processes. Can hold 60 Standard SBS Design plates or 25 Linbro plates.
Catalog No. NC2162981
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.