Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Torrey Pines Scientific™ EchoTherm™ Orbital Mixing Chilling/Heating Dry Bath

Used for any molecular biology lab mixes samples as they are heated or chilled with a variable speed orbital mixer

$8160.00 - $9180.00

Specifications

Certifications/Compliance UL, CSA, CE
Temperature Range -10°C to +100°C
Speed 200 to 1000rpm
Temperature Stability ±1°C
Width (English) 7.75 in.
View More Specs

Products 2
Catalog Number Mfr. No. Type Price Quantity & Availability  
Catalog Number Mfr. No. Type Price Quantity & Availability  
11-680-50
Accessories View Documents
Torrey Pines Scientific™
SC20
Non-programmable
Each for $8,160.00
Only null left
Add to Cart
 
11-680-51
Accessories View Documents
Torrey Pines Scientific™
SC25
Fully-programmable
Each for $9,180.00
Only null left
Add to Cart
 
Description

Description

EchoTherm™ SC20 Orbital Mixing Chilling/Heating Dry Bath is compact, versatile, reliable and easy to use. The built-in orbital mixer is designed with friction free hardened, blued, tempered steel leaf spring bearings that support the plate and sample blocks. Units come complete with instruction manual, universal bench top power supply for use anywhere in the world and AC line cord for the country of use. Sample blocks and covers are available as accessories.

  • Orbital mixing with speeds from 200 to 1000rpm
  • Energy-efficient Peltier design
  • Digital control to 1°C from -10° to 100°C
  • Control at room temperature
  • Thirty-day count down timer with audible alarm and user settable Auto-Off
  • RS-232 I/O port for control or data collection
  • Built-in data logger
  • Electronic calibration
  • Small compact size uses very little bench space
  • 12V DC operation. Universal power supply provided
Specification
  • Range: -10° to 100°C
  • Readability: 1°C
  • Accuracy:±1°C
  • Stability:±1°C
  • Sensor: Platinum RTD
  • Controller Type: PID
  • Uniformity:±0.5°C
    • Mixing samples in centrifuge tubes, assay plates, PCR plates, vials, test tubes or other containers while heating or chilling
    • Maintaining 14°C for ligation reaction
    • Maintaining 17°C for storing oocytes
    • Storing samples at ice bucket temperatures without the mess
    • Incubating samples at 37°C
    • Storing enzyme or DNA libraries at your workstation
    • Enzyme reactions and deactivations

    Compliance

    UL, CSA, and CE compliant

Specifications

Specifications

UL, CSA, CE
200 to 1000rpm
7.75 in.
4.25 in.
14.5 in.
24 lb.
100/265 V 50/60 Hz
50/60 Hz
-10°C to +100°C
±1°C
19.69 cm
10.8 cm
36.83 cm
10.9 kg
100/265 V
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.