Learn More
Zeptometrix Echovirus Type 13 (Strain: H06368 226) Culture Fluid
Shop All ZeptoMetrix Corporation Products
Description
Specifications
Specifications
| Biosafety Level | 2 |
| Certifications/Compliance | ISO 13485 |
| Product Type | Infectous Culture Fluid |
| For Use With (Application) | Nucleic Acid-Molecular Testing, Limit of Detection (LOD) Studies, Cross-reactivity Studies, Other Viral-based Assays |
| Quantity | 1 mL |
| Regulatory Status | RUO |
The purchase of infectious microorganisms from ZeptoMetrix requires a Material Transfer Agreement (MTA).