Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ECL Plex Goat anti-Mouse IgG-Cy™ 3 Secondary Antibody, Cytiva
GSA_VA

$358.05

Specifications

Antigen IgG
Classification Monoclonal
Conjugate Cyanine3
Form Purified
Host Species Goat
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
45-001-208
GSA_VA
View Documents
Cytiva
PA43009
150 μg
Each of 1 for $358.05
Only null left
Add to Cart
 
Description

Description

Host Species: Goat
Isotype: IgG
Species Reactivity: Ms

Western Blotting

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

IgG
Cyanine3
Goat
Secondary
Monoclonal
Purified
IgG
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.