Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ECL Plex Goat anti-Mouse IgG-Cy™ 5 Secondary Antibody, Cytiva

Catalog No. 45001210 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
150 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-001-210 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45-001-210 Supplier Cytiva Supplier No. PA45009
NLSU: Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Add to Cart

Host Species: Goat
Isotype: IgG
Species Reactivity: Ms

Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG
Classification Monoclonal
Conjugate Cyanine5
Host Species Goat
Quantity 150 μg
Primary or Secondary Secondary
Form Purified
Isotype IgG
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.