Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Eclipse Hypodermic Needles

Designed with pivoting needle technology for single-handed activation

Supplier:  BD 305166

 View more versions of this product

Catalog No. 14-821-13K



Due to product restrictions, please to purchase or view availability for this product.


Description

Description

No hard surface is required with this needle-based, safety-engineered injection device.

  • For use with BD Luer Lok
  • Sterile
  • Shelf Life: 5 years

Specifications

Specifications

Hypodermic Needle
Green
21 G
3.17 cm
Regular
Sterile
Yes
1.25 in.
Bevel Point
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit