Learn More
Celestron™ Eclipsmart™ Ultra 8 Piece Sun Observing and Imaging Kit
SCI_ED
Catalog No. S24542 Shop All Celestron International Products
Change view
Click to view available options
Eclipsmart™ Ultra 8 Piece Sun Observing and Imaging Kit
Catalog No.
S24542 Eclipsmart™ Ultra 8 Piece Sun Observing and Imaging Kit
1 options
Catalog No. S24542 Supplier Celestron™ Supplier No. 44414
All-in-one family pack that provides sun observing and solar eclipse viewing for up to six people.
  • Includes one pair solar glasses, four foldable paper solar shades, photo filter and 32-page book and bonus poster
  • Economical, easy to use and solar safe
  • Protection from harmful solar radiation, including both IR and UV light, and filters 99.99% of intense visible light
Specifications
Product Type Kit, Ultra 8 Piece Sun Observing and Imaging
Description Viewing for up to six people
For Use With (Application) Solar and eclipse viewing
Includes One clipsmart Solar Safe™ glasses, Four EclipSmart™ Solar Safe™ shades, One EclipSmart™ Solar Safe™ photo filter, One 32-page eclipse guide, One Eclipse transit map