Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ECM1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582960
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582960 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582960 Supplier Invitrogen™ Supplier No. PA582960
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EDTLDKYCDR EYAVKTHHHL CCRHPPSPTR DECFARRAPY PNYDRDILTI DIGRVTPNLM GHLCGNQRVL TKHKHIPGLI HNMTARCCDL PFPEQACCAE EEKLTFINDL CGPRRNIWRD PALCCYLSPG DEQVNCFNIN YLRNVALVS Highest antigen sequence indentity to the following orthologs: Mouse - 76%, Rat - 79%.

ECM1, also referred to as extracellular matrix protein 1, is a protein that plays a significant role in regulating extracellular matrix organization and cell adhesion. It is expressed in various tissues, including the skin, lung, and liver, and is involved in the development and progression of several diseases, including cancer and dermatological disorders. The protein interacts with several extracellular matrix proteins, such as collagen and fibronectin, and is involved in the regulation of cell proliferation, migration, and invasion. ECM1 is also involved in endochondral bone formation, angiogenesis, and tumor biology. It interacts with various extracellular and structural proteins, contributing to the maintenance of skin integrity and homeostasis. Mutations in this gene are associated with lipoid proteinosis disorder (also known as hyalinosis cutis et mucosae or Urbach-Wiethe disease) that is characterized by generalized thickening of skin, mucosae, and certain viscera. Different isoforms of ECM1 have been described in alternatively spliced transcript variants. Diseases associated with ECM1 include Lipoid Proteinosis Of Urbach And Wiethe and Lichen Sclerosus Et Atrophicus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ECM1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ECM1
Gene Accession No. Q16610
Gene Alias AI663821; ECM1; Extracellular matrix protein 1; LOW QUALITY PROTEIN: extracellular matrix protein 1; extracellular matrix protein 1; p85; RP11-54A4.6; Secretory component p85; testicular tissue protein Li 61; URBWD
Gene Symbols ECM1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ECM1. Recombinant protein control fragment (Product #RP-89165).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1893
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.