Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ECM29 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA3035 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA3035 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA3035 Supplier Invitrogen™ Supplier No. PA3035
  • $648.90
    $500.65 / Each of 1

    Save $148.25

    23% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

PA3-035 detects ECM29 in human and mouse samples. PA3-035 has been used successfully in Western blot and immunofluorescence procedures. In Western blot analysis of HeLa cell extracts this antibody detects a ~205 kDa protein representing ECM29. The PA3-035 immunogen is a synthetic peptide corresponding to the C-terminal residues R(1821) P E L Q E K A A L L K K T L E N L E(1839) of human ECM29.

ECM29 appears to be one of three proteins which associate significantly with proteasomes. ECM29 has been shown to enhance the stability of the proteasome. ECM29 deletion has been shown to produce 26S proteasomes which dissociated in the absence of ATP. It was thus suggested that ECM29 tethers the regulatory complex to the proteasome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ECM29
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation whole serum with 0.05% sodium azide
Gene ECPAS
Gene Accession No. Q5VYK3, Q6PDI5
Gene Alias AI314180; AW558785; BB181316; ECM29; ECM29 homolog, proteasome accessory protein; Ecpas; expressed sequence AI314180; FLJ22036; homolog of yeast ECM29; KIAA0368; KIAA1962; mKIAA0368; Proteasome adapter and scaffold protein ECM29; Proteasome-associated protein ECM29 homolog; RP11-386D8.2
Gene Symbols AI314180, ECPAS
Host Species Rabbit
Immunogen Synthetic peptide corresponding to the C-terminal residues R(1821) P E L Q E K A A L L K K T L E N L E(1839) of human ECM29.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 230249, 23392
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.